Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TerraCycle™ Used Chewing Gum Zero Waste Box™

Recycle Used Chewing Gum. Each box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.
$118.40 - $240.00
Specifications
Color | Brown, Green, White |
---|---|
Type | Recycling Box |
Material | Cardboard |
Disposable | Yes |
Latex Free | Yes |
Alert:
Not available in Hawaii, Alaska, and Puerto Rico.
Catalog Number | Mfr. No. | Capacity (Metric) | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
S88854141
![]() |
TerraCycle™
CS39S |
39.66 L | 11 x 11 x 20 in. |
Each for $118.40
|
|
S88854142
![]() |
TerraCycle™
CS39M |
79.31 L | 11 x 11 x 40 in. |
Each for $157.40
|
|
S88854143
![]() |
TerraCycle™
CS39L |
136.42 L | 15 x 15 x 37 in. |
Each for $240.00
|
|
Description
- WHAT YOU CAN RECYCLE
- Please send us used chewing gum.
- Do not send in other types of food or candy.
- How It Works
- Order: Select a Zero Waste Box system and place your order.
- Collect: Collect the accepted items to fill the system.
- Ship: Send it back to TerraCycle with the prepaid return label on your system.*
- Reorder: Order your next Zero Waste Box system to continue to recycle everything.
- What’s Included?
- One Zero Waste Box
- One prepaid return label, affixed to your box
- Processing and recycling of the materials
- Customer support from our team of experts
- Important Shipping Restrictions
- For the Zero Waste Box system to work effectively and safely, please follow these simple shipping guidelines on how and what you can ship back to TerraCycle:
- Only send in items this system accepts. Sending in items that fall outside the selected category causes contamination and the need for added separation, which increases costs.
- Do not send in household items that may be considered hazardous waste. These include items such as lithium-ion batteries, pressurized canisters, pesticides, oil-based paint or medical sharps, organics, broken glass, biomedical waste, soiled diapers, or expired medication. Do not send in anything sharp, flammable, reactive, corrosive, ignitable, radioactive, toxic, infectious, or pathogenic.
- Do not send in medical waste. This includes material contaminated with blood or bodily fluids that originate from health care facilities, hospitals, physicians offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
- Do not ship dripping packages. Please make sure all excess product has been removed before putting it in your Zero Waste Box, Pouch, or Pallet. If you choose to rinse your items, please note that the system must be completely dry prior to shipping.
- For the Zero Waste Box system to work effectively and safely, please follow these simple shipping guidelines on how and what you can ship back to TerraCycle:
Specifications
Brown, Green, White | |
Cardboard | |
Yes | |
Not Autoclavable | |
Plastic |
Recycling Box | |
Yes | |
Non-sterile | |
Cardboard Box | |
Recycling |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title