Learn More
Grainger FIRST AID ONLY, Fire Blanket, Fire Blanket
Supplier: Grainger 39P028
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The First Aid Only Fire Blanket is a flame retardant wool blanket designed to smother small fires. It measures 60" x 80" and is made of 100% wool with vermiculite insulation between two layers. The fire blanket extinguishes fires by cutting off oxygen supply. It can be used on stovetop, electrical, grease and oil fires. The fire blanket comes in a plastic bag with handles for easy storage and access. It meets the EN 1869 European standard for non-flammability. This fire blanket provides an easy to use option to fight small fires in the kitchen, workshop or campsite.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.