Learn More
Carbosynth LLC. Urokinase-Derived Peptide A6, 1mg, MF= C39H62N10O15
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Carbosynth LLC. UPA3038PI1MG
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This item has a minimum order quantity of 4 per supplier requirements
Ac-Lys-Pro-Ser-Ser-Pro-Pro-Glu-Glu-NH2; A6 Peptide; Urokinase Plasminogen Activator (136-143), human; Acetyl-Lysyl-Prolyl-Seryl-Seryl-Prolyl-Prolyl-Glutamyl-Glutamic Acid Amide; uPA-Derived Peptide A6, Application: CD44 Inhibitor; Anti-Tumor Properties, Notes: Sequence derived from peptide domain of human urokinase plasminogen activator (uPA). It is able to bind to and inhibit CD44 activity that contributes to tumorgenesis., salt: Trifluoroacetate Salt, one letter code: Ac-KPSSPPEE-NH2, MW: 910.99
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.