Learn More
Quantimetrix Dropper Urinalysis Dipstick Control, Level 1&2
SDP
Supplier:  Quantimetrix 144002
Encompass_Preferred
•Easy-to-use dropper bottles•Liquid, made with human urine•24-month shelf life & open vial stability from date of manufacture when stored at 2ºC–8ºCMonitor the performance of visual and instrument readings of urine dipsticks by dropping the control directly onto the dipstick. Features an easy-to-use dropper bottle for simple dispensing and maximum use. We include values for all dipstick analytes plus microalbumin and creatinine, and qualitative results for hCG early pregnancy detection test methods. Dropper Urinalysis Dipstick Control is designed for use with most urinalysis reagent strips. It can also be used for confirmatory tests and refractometry.Thoughtfully redesigned packaging now includes barcodes on the labels for lot numbers and expiration dates and we’ve made it easier to identify each level.Dropper Analytes: Bilirubin, Blood, Creatinine, Glucose, hCG, Ketones, Leukocytes, Microalbumin, Nitrite, pH, Protein, Specific Gravity, Urobilinogen
Catalog No. NC9667223
Explore available promotions