Learn More
Fisherbrand™ Urine Analyzer for Urinalysis Reagent Strips, CLIA Waived
Urine Analyzer, for use with Urinalysis Reagent Strips for the semi-quantitative detection of the following analytes in urine: Glucose, Bilirubin, Ketone (Acetoacetic acid), Specific Gravity, pH, Blood, Protein, Urobilinogen, Leukocytes, Nitrites.
Description
A CLIA Waived, easy-to-use system that provides fast, highly-accurate results for the general evaluation of health, aiding in the diagnosis and monitoring of metabolic or systemic diseases that affect kidney function, endocrine disorders and diseases or disorders of the urinary tract.
Features:
- Strips combinations with up to 10 parameters
- Parameters available include microalbumin, and creatinine.
- Analytical sensitivity comparable to the market leader
- High quality color chart ensures accurate visual reading
- 2 year shelf life for unopened canisters
- 3 month shelf life for strips in opened canisters
High Accuracy and Reliability:
- Automatic calibration before each test
- Up to 120 tests per hour in Continuous Mode
- Reads strips with up to 10 parameters, including
- Microalbumin/Creatinine
- Minimal training required
- Robust construction
- Data Management ready
- Includes RS232C and USB ports for data transfer to an external computer or LIS
- 510(k) Cleared and CLIA Waived
Specifications
Urine Analyzer | |
English, French, Spanish, Italian, Portuguese | |
Automatically saves the last 2,000 results | |
Analyzer: 27.5 x 18.3 x 13.9 cm (10.8 x 7.2 x 5.5 in.); Display Screen: 10.8 x 10.8 x 6.5 cm (4.2 x 4.2 x 2.6 in.) | |
50/60 Hz |
LCD | |
Strip Incubation Time: 1 min. | |
Single Test Mode: 60 Tests/hr.; Continuous Test Mode: 120 Tests/hr. | |
1 Unit | |
100/240 V |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.