Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Urea, Ultrapure, 99.5%, Spectrum™ Chemical
Click to view available options
Quantity:
25 g
125 g
500 g
1 kg
5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Urea, Ultrapure is an odorless and colorless solid that is an important nitrogen-containing substance found in mammal urine. Urea is an effective protein denaturant. In a highly concentrated (6 to 8 M) urea solutions, many proteins unfold and separate into their constituent polypeptide chains. Urea UltraPure is also used to denature nucleic acids in sequencing gels. Ultrapure reagents offered by Spectrum undergo rigorous specification testing to insure accurate, repeatable results.
Specifications
CAS Max % | 1 |
Melting Point | 132°C to 135°C |
Packaging | Poly Pail |
Quantity | 12 kg |
Percent Purity | 99.5% |
Grade | Ultrapure |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction