Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Uranyl Nitrate 5 GR
SDP

Supplier:  Electron Microscopy Sciences 22600

Encompass_Preferred

CAS #13520-83-7 UO₂(NO₃)2·₆H₂O F.W. 502.13
Solubility: ~66g/100g H2O
Melting point: 60°C
Boiling point: 118°C

A Universal EM Negative Stain used for viruses. In tissue samples it stabilizes nucleic acids and cell membranes. These solutions are more stable than uranyl acetate and they react primarily with negatively charged groups in the absence of phosphate ions. Reagent Grade, A.C.S. UO2(NO3)2·6H2O

Catalog No. 50-189-8761

This product is regulated by the federal government, and may not be ordered online.


Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit