Learn More
Electron Microscopy Sciences Uranyl Magnesium Acetate 1 GR
Supplier: Electron Microscopy Sciences 22501
CAS #20596-93-4 UO₂Mg(CH₃COO)₄ F.W. 530.0
Used as a substitute for uranyl acetate in either double-staining techniques and also as a solo stain.