Learn More
Ted Pella Inc Uranyl Acetate, 6159-44-0, MFCD00150376, 25g
Supplier: Ted Pella Inc 19481
Molecular Formula C4H6O6U.2H2O, Molecular Weight 424.145, Positive or negative stain, EM. May cause adverse mutagenic or teratogenic effects.