Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Upper Triple Root Molar with Caries Model

Enlarged 15 times to show the finest details.
Supplier: Eisco™ AM0046
Description
- Hand painted molar model
- Separated into two longitudinal sections through crown, two roots and pulp cavity
- Mounted on base
- Supplied with English key card
Specifications
Model, Molar | |
Shows upper triple root molar with caries | |
Base |
13 x 13 x 21 cm | |
English key card | |
300 g |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title