Learn More
Idex Stainless-Steel Sample Loop
Compatible with Rheodyne™ analytical injection valves
Supplier:  Idex 1876
Encompass
Catalog No. 05-701-235
Add to Cart
Description
Description
Leads are long enough to be used at 40° and 60°. For Rheodyne Models 7125, 7010 (leads angled at 40°); and Models 7725, 7725i (leads angled at 60°).
  • Fits into Rheodyne analytical sample injection valves, with leads conveniently bent at appropriate angles to allow trouble-free installation
  • For use with Rheodyne valve models 7125, 7010
  • Theoretical volumes are consistently controlled with use of electrochemical cutting machine (ECM)
  • Each loop passivated and flushed with reagent-grade methanol
  • Fittings and adapters accompanying sample loops rated to 1000psi (69 bar)
  • Volumes stated at ±10% with exact calibration
Each calibrated sample loop is documented and supplied with calibration certificate.
Specifications
Specifications
Sample Loop
Analytical Instrumentation, in-Vitro Diagnostics, Biotechnology, Laboratory
0.2 cm
Stainless Steel
0.08 in.
10 mL
Product Suggestions
Product Suggestions