Learn More
OpGen Unyvero L4 Lysator Dust Filter Fleece
For use with Unyvero L4 Lysator (60002).
Supplier: OpGen 60008
Alert:
Filter needs to be replaced once a year or more frequently depending on the amount of airborne particles such as dust in the surrounding environment. Also replace Unyvero A50 Analyzer Dust Filter Fleece (60007) and Unyvero A50 Analyzer Plug-In Filter 2x (60009) at the same time.
Description
- Unyvero L4 Lysator Dust Filter Fleece (60008) is the replacement filter for use with Unyvero Lysator (60002)
- 1 filter per box
- Filter needs to be replaced once a year or more frequently depending on the amount of airborne particles such as dust in the surrounding environment; also replace Unyvero A50 Analyzer Dust Filter Fleece (60007) and Unyvero A50 Analyzer Plug-In Filter 2x (60009) at the same time.
Specifications
Replacement Filter | |
End-point PCR |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Filter needs to be replaced once a year or more frequently depending on the amount of airborne particles such as dust in the surrounding environment. Also replace Unyvero A50 Analyzer Dust Filter Fleece (60007) and Unyvero A50 Analyzer Plug-In Filter 2x (60009) at the same time.