Learn More
OpGen Unyvero A50 Analyzer Dust Filter Fleece
For use with Unyvero A50 Analyzer (60001).
Supplier: OpGen 60007
Alert:
Filters need to be replaced once a year or more frequently depending on the amount of airborne particles such as dust in the surrounding environment. Also replace Unyvero A50 Analyzer Plug-In Filter 2x (60009) and Unyvero L4 Lysator Dust Filter Fleece (60008) at the same time.
Description
- 2 filters per box
- Filters need to be replaced once a year or more frequently depending on the amount of airborne particles such as dust in the surrounding environment; also replace Unyvero A50 Analyzer Plug-In Filter 2x (60009) and Unyvero L4 Lysator Dust Filter Fleece (60008) at the same time
Specifications
Replacement Filter | |
End-point PCR |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Filters need to be replaced once a year or more frequently depending on the amount of airborne particles such as dust in the surrounding environment. Also replace Unyvero A50 Analyzer Plug-In Filter 2x (60009) and Unyvero L4 Lysator Dust Filter Fleece (60008) at the same time.