Learn More
Cole-Parmer Koflo 3/16-12P Unmounted in-line mixer; 3/16"OD, 12 elements, 10/pack
Supplier:  Cole-Parmer 0466702
Encompass_Preferred
Unmounted mixers can be used with any standard tubing having the same tubing ID as the element OD given in the table. They work well with flexible tubing.
Catalog No. NC0799272