Learn More
Newcomer Supply Microtome Aligner - Universal. Align your microtome in minutes!
Supplier: Newcomer Supply 6257TO
A simple leveling tool that can align your microtome in the X, Y and Z axis in only a few minutes. Very easy to use and will fit on all microtomes. Every microtome can be ready to do recuts on those precious biopsies! So simple & quick to use. Only one Universal Microtome Aligner tool needed for ALL models.