Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Poltex Universal Locking Wipe Cannister Holder

Holds virtually any wipe cannister, mask box, glove box, tissue box, etc.
$89.10 - $107.80
Catalog Number | Mfr. No. | Mounting | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Mounting | Price | Quantity & Availability | |||||
22-000-514
![]() |
Poltex
MINMAX59M |
Magnet mount |
Each for $107.80
|
|
22-000-515
![]() |
Poltex
MINMAX59W |
Wall mount |
Each for $89.10
|
|
Description
- Locks securely with padlock (included)
- Comes with powerful magnets for mounting
- Inside height adjusts in 0.5 in. increments from 5 to 9 in.
- Choose from keyhole slots for wall (or countertop) mount (included)
- Constructed from durable and cleanable 0.25 in. thick PETG
- Holds virtually and wipe cannister, mask box, glove box, tissue box, wipe packet, etc.
- Consumables not included
Specifications
Dispenser | |
Adjustable, Lockable |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title