Learn More
Simport™ Scientific Unisette™ Tissue Processing/Embedding Cassettes in Quickload™ Stacks
Unisette™ Tissue Processing/Embedding Cassettes in Quickload™ Stacks is offered with a 45° anterior writing surface. Configuration is considered ideal for optimal workflow in some automated cassette printers and efficient barcode reading.
$201.00 - $208.00
Specifications
Quantity | 1000/Cs. |
---|---|
Writing Face Angle | 45° |
Material | Acetal |
Product Type | Cassette |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
23-045-551
|
Simport™ Scientific
M52511T |
Orange |
Case of 1000 for $208.00
|
|
|||||
23-045-552
|
Simport™ Scientific
M52512T |
Aqua |
Case of 1000 for $201.00
|
|
|||||
23-045-553
|
Simport™ Scientific
M5252T |
White |
Case of 1000 for $208.00
|
|
|||||
23-045-554
|
Simport™ Scientific
M5253T |
Pink |
Case of 1000 for $201.00
|
|
|||||
23-045-555
|
Simport™ Scientific
M5254T |
Green |
Case of 1000 for $208.00
|
|
|||||
23-045-556
|
Simport™ Scientific
M5255T |
Yellow |
Case of 1000 for $201.00
|
|
|||||
23-045-557
|
Simport™ Scientific
M5256T |
Blue |
Case of 1000 for $201.00
|
|
|||||
23-045-558
|
Simport™ Scientific
M5257T |
Peach |
Case of 1000 for $201.00
|
|
|||||
23-045-559
|
Simport™ Scientific
M5258T |
Tan |
Case of 1000 for $201.00
|
|
|||||
23-045-560
|
Simport™ Scientific
M5259T |
Gray |
Case of 1000 for $201.00
|
|
|||||
Description
- Keeps specimens safely submerged in liquid and are resistant to the chemical action of most histological solvents.
- Efficient flow-through slots maximize fluid exchange and ensure proper drainage.
- One-piece construction with snap-latch and hinge-lock design allows for simple one-hand operation and prevents early separation of the base and lid.
- Can be opened and closed as often as necessary and they re-lock securely without the danger of specimen loss.
- Convenient Quickload™ stacks are suitable for automated printers.
Specifications
1000/Cs. | |
45° | |
Acetal | |
Cassette |