Learn More
Simport™ Scientific Unisette™ Biopsy Processing/Embedding Cassettes in Quickload™ Stacks
Secure specimens during processing with Unisette™ Biopsy Processing/Embedding Cassettes in Quickload™ Stacks. The snap latch and hinge-lock design of the UNISETTE™ allow for one-hand operation and always relock securely without danger of specimen loss.
$366.00 - $377.00
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
22-038-078
|
Simport™ Scientific
M5062T |
White |
Case of 1000 for $366.00
|
|
|||||
22-038-079
|
Simport™ Scientific
M5063T |
Pink |
Case of 1000 for $366.00
|
|
|||||
22-038-080
|
Simport™ Scientific
M5064T |
Green |
Case of 1000 for $366.00
|
|
|||||
22-038-081
|
Simport™ Scientific
M5065T |
Yellow |
Case of 1000 for $366.00
|
|
|||||
22-038-082
|
Simport™ Scientific
M5066T |
Blue |
Case of 1000 for $377.00
|
|
|||||
22-038-512
|
Simport™ Scientific
M5067T |
Peach |
Case of 1000 for $366.00
|
|
|||||
22-038-513
|
Simport™ Scientific
M5068T |
Tan |
Case of 1000 for $366.00
|
|
|||||
22-038-514
|
Simport™ Scientific
M5069T |
Gray |
Case of 1000 for $366.00
|
|
|||||
22-038-515
|
Simport™ Scientific
M50610T |
Lilac |
Case of 1000 for $366.00
|
|
|||||
22-038-516
|
Simport™ Scientific
M50611T |
Orange |
Case of 1000 for $366.00
|
|
|||||
22-038-517
|
Simport™ Scientific
M50612T |
Aqua |
Case of 1000 for $366.00
|
|
|||||
Description
- In Quickload™ stacks
- 35° Anterior writing area
- One-piece construction with hinge-lock design prevent early separation of base and lid
- Snap latch and hinge-lock design - cassettes will always relock securely
- Allows for one-hand operation
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
Order Info
Each case contains 25 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
1000/Pk. | |
Acetal |