Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific™ UniSafe™ Vented Wash Bottle
Click to view available options
Chemical Label:
Acetone
Acetonitrile
Dichloromethane
Distilled Water
Ethanol
Ethyl Acetate
Isopropanol
Methanol
Methyl Ethyl Ketone
Sodium Hypochlorite
Toluene
Water
Description
Pre-printed with the Chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment. Each bottle is compliant with GHS and OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200. Intended for use only with LDPE and PP compatible solvents.
- Industrial quality low density polyethylene (LDPE) construction with long lasting strength. GHS compliant
- Preprinted bottles include: chemical name and formula, NFPA diamond, health hazards, and CAS number
- Polypropylene stem closure and draw tube
- Safety vented and color coded for quick solvent recognition
- 4-color printing, 500 mL (16 oz.) capacity
Specifications
Closure Material | Polypropylene |
Closure Type | Vented Screw Cap |
Through the Closure Dispensing | Yes |
Chemical Label | Acetonitrile |
Product Type | Vented Wash Bottle |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Yellow |
For Use With (Application) | LDPE and PP-compatible Solvents |
Material | LDPE |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction