Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific™ UniSafe™ Vented Wash Bottle, 1000 mL
Click to view available options
Chemical Label:
Acetone
Distilled Water
Ethanol
Isopropanol
Methanol
Sodium Hypochlorite
Description
- United Scientific™ UniSafe™ vented wash bottles are made from industrial grade low-density polyethylene with polypropylene stem closure and venting assembly.
- They are pre-printed with the Chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment.
- Each bottle is compliant with GHS and OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.
- Intended for use only with LDPE and PP compatible solvents.
- Industrial quality low density polyethylene (LDPE) construction with long lasting strength. GHS compliant
- Polypropylene stem closure and draw tube
- Safety vented and color coded for quick solvent recognition
- 4-color printing, 1000 mL capacity
- Available in 6 different varieties.
- Sold as packs of 4 bottles.
Specifications
Product Type | Safety-vented Wash Bottle |
Chemical Label | Acetone |
Capacity (Metric) | 1000 mL |
Capacity (English) | 33.8 oz. |
Color | Translucent |
Material | LDPE |
Sterility | Non-sterile |
Closure Included | Yes |
Autoclavable | Not Autoclavable |
Disposable | Reusable |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction