Learn More
Perkin Elmer US LLC Un-Assembled Aluminum Crimp Cap with PTFE/Red Rubber Septa and Star Spring, 100/pk
Supplier: Perkin Elmer US LLC N9302979
For sealing sample vials for temperatures up to 160°C. Recommend for storage and for multiple injection use.