Learn More
Epredia™ UltraVision Quanto Detection System AP
Delivers higher lab profitability and better staining by supporting laboratories to select the best antibody for each test.
Supplier: Epredia™ TL060QAL
Description
UltraVision Quanto AP has been validated in combination with Liquid Fast-Red Substrate System and provides four major benefits:High sensitivity—can both be used for tricky and challenging markers and/or allows a more gentle HIER
Increased sensitivity for rabbit antibodies—more economical use of expensive rabbit monoclonal antibodies Crisp, clear and strong staining for high contrast
Reduced incubation times for shorter run times&bulletc1_end; UltraVision Quanto is the technology of this decade in polymeric labeling. Polymer detection methods have been shown to provide increased sensitivity and detection simplicity. This innovative micropolymer technology has major advantages over conventional IHC systems as it allows the “detector” to readily penetrate to the nuclear membrane enabling the user to detect nuclear antigens without sacrificing the ability to stain cytoplasmic and membrane antigens. UltraVision Quanto amplifies the signal with both mouse and rabbit primary antibodies making it versatile and easy to use. Background noise due to nonspecific binding to endogenous biotin molecules is eliminated because UltraVision Quanto is not a biotin/avidin based system. The signal to noise ratio is further enhanced to detect low antigen expression or compensate for low primary antibody affinity by utilizing highly purified secondary antibodies and enzymes. The UltraVision Quanto ultimately provides the user with a rapid, easy to use, and versatile IHC detection system that offers superior staining of tissues traditionally thought difficult to visualize.
Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Enzyme: Alkaline Phosphatase
Chromogen/Substrate: None provided
Includes: TL-060-AL: Primary Antibody Amplifier Quanto (TL-060-QPB), AP Polymer Quanto (TL-060-QAP), Ultra V Block (TA-060-UB)
TL-125-AL: Primary Antibody Amplifier Quanto (TL-125-QPB), AP Polymer Quanto (TL-125-QAP), Ultra V Block (TA-125-UB)
Specifications
None provided | |
Lab Vision™, UltraVision™ | |
None provided | |
60mL |
Alkaline Phosphatase | |
anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L) | |
Primary Antibody Amplifier Quanto (TL-060-QPB), AP Polymer Quanto (TL-060-QAP), Ultra V Block (TA-060-UB) |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.