Learn More
Epredia™ UltraVision LP HRP Polymer & DAB Detection System
Epredia™ Lab Vision UltraVision LP Detection System: HRP Polymer/DAB Plus Chromogen is a sensitive detection system based on biotin-free, polymer technology.
Supplier: Epredia™ TL015HD
Description
UltraVision LP is the latest technology in polymeric labeling. Polymer detection methods have been shown to provide increased sensitivity and detection simplicity. This second-generation polymer system is composed of smaller polymer subunits that minimize conflicts in binding the target protein. Decreased binding conflicts result in more consistent staining and better signal amplification.1 Ultimately, this gives the user higher sensitivity and antibody efficiency.2 With UltraVision LP, you use less antibody and obtain better signal-to-noise ratios. UltraVision LP is also biotin-free, which eliminates background staining found with traditional biotin-based detection methods.Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Enzyme: Peroxidase
Chromogen/Substrate: Diaminobenzidine (DAB)
Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Enzyme: Peroxidase
Chromogen/Substrate: Diaminobenzidine (DAB)
Includes:
TL-015-HD: 15mL Hydrogen Peroxide Block (TA-015-HP), 15mL Ultra V Block (TA-015-UB), 15mL Primary Antibody Enhancer (TL-015-PB), 15mL HRP Polymer (TL-015-PH), 15mL DAB Plus Substrate (TA-015-HSX), 1mL DAB Plus Chromogen (TA-001-HCX)
TL-060-HD: 60mL Hydrogen Peroxide Block (TA-060-HP), 60mL Ultra V Block (TA-060-UB), 60mL Primary Antibody Enhancer (TL-060-PB), 60mL HRP Polymer (TL-060-PH), 60mL DAB Plus Substrate (TA-060-HSX), 2mL DAB Plus Chromogen (TA-002-HCX)
TL-125-HD: 125mL Hydrogen Peroxide Block (TA-125-HP), 125mL Ultra V Block (TA-125-UB), 125mL Primary Antibody Enhancer (TL-125-PB), 125mL HRP Polymer (TL-125-PH), 125mL DAB Plus Substrate (TA-125-HSX), 4mL DAB Plus Chromogen (TA-004-HCX)
Note:
UltraVision detection system detects a specific mouse IgG or rabbit IgG antibody bound to an antigen in tissue sections. The specific antibody is located by a universal secondary antibody formulation conjugated to an enzyme-labeled polymer that recognizes mouse and rabbit immunoglobulins. The polymer complex is then visualized with an appropriate substrate/chromogen.
Specifications
150 to 300 Slides | |
15mL Hydrogen Peroxide Block (TA-015-HP), 15mL Ultra V Block (TA-015-UB), 15mL Primary Antibody Enhancer (TL-015-PB), 15mL HRP Polymer (TL-015-PH), 15mL DAB Plus Substrate (TA-015-HSX), 1mL DAB Plus Chromogen (TA-001-HCX) | |
anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L) | |
Peroxidase |
15mL | |
Diaminobenzidine (DAB) | |
Store at 2° to 8°C. Each component is stable for 18 months. | |
Diaminobenzidine (DAB) |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.