Learn More
Ted Pella Inc Ultrathin Carbon Film on Lacey Carbon Support Film, 300 mesh, Gold, 25/pk
Supplier: Ted Pella Inc 01824G
Pure carbon support films with no Formvar backing. This new product has an even thinner carbon film which is mounted on a Lacey carbon film. The carbon support layer lying over the holes is less than 3nm in thickness and is the thinnest support film available. It is particularly useful for high resolution microscopy of low contrast particles and is also ideal for use with the Energy Filtering TEM.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.