Learn More
Cole-Parmer Cole-Parmer Ultrasonic Cleaner, Heater/Digital Timer; 0.5 gal, 115V
Supplier: Cole-Parmer 0889527
These cleaners transform low-frequency AC current into 40 kHz high-frequency sound waves using a piezoelectric transducer. The transducer creates sinusoidal waves, which in turn cause cavitationthe formation and violent collapse of minute vacuum bubbles in the solution. These implosions thoroughly scrub every surface with which the solution makes contact, yet are not harsh on delicate items. Cleaners also feature sweep frequency that provides uniform cleaning throughout the tank by creating overlapping ultrasonic waves and eliminating inconsistent cleaning due to areas of intense ultrasonic activity.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.