Learn More
Innovative Med Tech UltraFlo-L Safety Butterfly Blood Collection Set, 23G x 7 in.
The UltraFlo-L Safety Butterfly Blood Collection Set utilizes the safety-shield activation technique with proprietary bevel technology which requires lower penetration force for an improved draw experience.
Supplier: Innovative Med Tech 292
Description
- Affordability without any change in technique
- Utilizes proven safety-shield activation technique
- 7 in. tubing
UltraFlo™-L Safety Blood Collection Set with UltraFlo Cannula (23G, 3/4 x 7 in.)
- Increase 23G flow rate 200%*
- Improve patient comfort - use 23G instead of 21G without compromising flow rate
- Proprietary bevel technology requires lower penetration force* for an improved draw experience
- *3rd-party testing compared to leading brand
UltraFlo-L™ Safety Shield
- Same design, no change in technique
- One or two-handed activation
Specifications
Blood Collection Set | |
PVC | |
Sterile | |
7 in. |
Blue | |
23 G | |
Yes |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.