Learn More
World Precision Instrument UltraDose Enzyme Plus Ultrasonic Cleaning Solution
SDP
Supplier:  World Precision Instrument WPI038
Encompass_Preferred
This EPA approved Super concentrate solution cleans and disinfects non-porous instruments and apparatus surfaces in a 10-minute period. Eliminates residual micro-organisms in a single step. Performs as an ultrasonic cleaner, disinfectant, deodorant, germicidal, fungicidal, viricidal and tuberculocidal all in one. Works well on MRSA.  To be used in an ultrasonic cleaner.
Catalog No. 50-253-8257