Learn More
Perkin Elmer US LLC Ultra Quiet Compact Oil-Free Compressor - 220 V, 50 Hz
Supplier: Perkin Elmer US LLC N9306351
This compressor features a built-in dessicant dryer, providing clean, dry, and particle-free air at -40 dew point. It is excellent for gas chromatography applications involving air generators. It is commercial/industrial grade, ultra quiet for use in the laboratory area and contains built-in rolling casters and a convinient handle. It comes with a 1 year warranty on workmanship. Power Requirements: 220 V/50 Hz Horsepower: 0.75 Output: 4.4 CFM/110 L/min Operating Pressure: 80-100 psi/6-7 bar Maximum Pressure: 100 psi/7 bar Noise Level: 57 db/A Tank Size: 6 L/1.57 Gal Dimensions: 34.7 in x 34.7 in x 22.4 in (Packed: 38 in x 15 in x 25 in) Weight: 103 lbs (Packed: 125 lbs)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.