Learn More
Perkin Elmer US LLC Ultra Low Particulate V-Solv, 5 Gal
Supplier:  Perkin Elmer US LLC N9308750
Encompass_Preferred
Like regular V-Solv, our Ultra-Low Particulate V-Solv is designed for chemical stability of metals in oil and maintains performance in a wider range of environmental conditions. Ultra Low Particulate V-Solv + Cobalt has the same low level of particulates as ULP V-Solv but has Cobalt premixed at 40 µg/g for use as a dilution solvent for the hyphenated particle counter configuration. Matrix: Volume: Content: Ultra Low Particulate V-Solv 5 Gal Cobalt
Catalog No. 50-211-3248