Learn More
Perkin Elmer US LLC Ultra Low Particulate V-Solv - Colbalt, 5 Gal
Supplier:  Perkin Elmer US LLC N9308749
Encompass_Preferred
Like regular V-Solv, our Ultra-Low Particulate V-Solv is designed for chemical stability of metals in oil and maintains performance in a wider range of environmental conditions. Ultra Low Particulate V-Solv provides a consistently low background of particles for dilution solvent for standalone Particle Counter configuration and rinse for both the standalone and hyphenated particle counting configurations. Matrix: Volume: Content: V-Solv 5 Gal Blank
Catalog No. 50-211-3249