Learn More
Leica Biosystems ULTRA FINE MARKING PENS 12 EA
Supplier: Leica Biosystems 3801880
Marking Pens are extremely versatile and can be used on various writing surfaces found in the laboratory. They have an extra fine point made of rigid nylon for precise marking on glass slides as well as tissue cassettes. The quick drying ink is resistant to most aqueous based stains, alcohols, xylene and xylene substitutes. A tight fitting cap helps prevent drying of the tip. The pen barrel is ergonomically designed for ease of use.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.