Learn More
Vector Laboratories Ulex Europaeus Agglutinin I (UEA I) Rhodamine-labeled 2 mg
Supplier: Vector Laboratories RL10622
Ulex europaeus agglutinin I (UAE I) binds to glycoproteins and glycolipids containing α-linked fucose residues, such as ABO blood group glycoconjugates. This lectin preferentially binds blood group O cells and has been used to determine secretor status. It is also an established marker for human endothelial cells. This rhodamine-labeled UAE I features a ratio of fluorophore to lectin protein that provides optimal staining (excitation 575 nm, emission 600 nm) and is supplied in solution, essentially free of unconjugated fluorophore.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.