Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCR Safety Matrix Headgear
Click to view available options
Matrix Crown Face Shield
Description
Offers superior protection from liquid splashes and flying debris that would otherwise be damaging to the face
Compliance
Meets ANSI Z87.1+ and CSA Z94.3 standards.
Specifications
Certifications/Compliance | ANSI Z87.1+, CSA Z94.1 |
Lens Tint | Clear |
For Use With (Equipment) | Matrix headgear |
Lens Material | Polycarbonate |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction