Learn More
DuPont™ Tyvek™ 400D Pants
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
DuPont™ Tyvek™ 400 D pants have been designed for very specific applications where demanding comfort requirements are combined with limited protective requirements for frontal exposures. They are well suited for workers who are involved in a variety of strenuous activities that can lead to heat stress in applications that include:
- Wind Turbines
- Composites manufacturing
- Boat manufacturing
- Remediation
- Utilities
- Maintenance
- Glass manufacturing
Specifications
Size | X-Large |
Material | Spunbond Polypropylene |
Color | White |