Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ Tyvek™ 400 Disposable Lab Coats, USMCA Sourced
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
DuPont™ Tyvek™ 400 provides lightweight inherent barrier protection against hazardous dry particles and aerosols, and nonhazardous light liquid splash
- Abrasion resistance due its unique, nonwoven material
- Can be used in a variety of applications including general maintenance, operations, asbestos and lead abatement, mold remediation, and environmental cleanups
Compliance
NAFTA
Specifications
Specifications
Product Type | General Purpose Lab Coat |
Size | Medium |
Gender | Unisex |
Material | Tyvek |
Color | White |
Closure Type | Snaps |
Disposable | Limited-use |
Sterility | Non-sterile |
No. of Pockets | 2 |
Model | TY212SWH |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction