Learn More
DuPont™ Tyvek™ 400 Comfort Fit Design Disposable Coverall with Collar, Elastic Wrists and Ankles, NAFTA
GREENER_CHOICE
Catalog Number 17500219
This item is not returnable. View return policy
This item is not returnable. View return policy
DuPont™ Tyvek™ 400 coveralls with elastic wrists and ankles provide the protection and durability you need so you can focus on the task at hand. Meets NAFTA Standards.
DuPont™ Tyvek™ 400 provides light-weight inherent barrier protection against hazardous dry particles and aerosols, and nonhazardous light liquid splash.
  • Excellent abrasion resistance due its unique, nonwoven material
  • Can be used in a variety of applications including general maintenance/operations, asbestos and lead abatement, mold remediation, and environmental cleanups
Compliance
NAFTA