Learn More
CPC Scientific H-Tyr-Gln-Glu-Ala-Phe-Arg-Arg-Phe-Phe-Gly-Pro-Val-OH (trifluoroacetate salt) 5MG
Supplier: CPC Scientific OSTP003B
Osteocalcin analog.ONE-LETTER SEQUENCE: YQEAFRRFFGPVMOLECULAR FORMULA: C73H101N19O17MOLECULAR WEIGHT:1516.7STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [129356-77-0], RESEARCH AREA: OsteoporosisREFERENCES: J. Podenphant et al., Calcif. Tissue Int., 36, 536 (1984)