Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Accuform™ Type I Plastic Barricade
Accuform™ Type I Plastic Barricade are heavy duty, and highly visible to warn of hazard situations in low light conditions.
Supplier: Accuform™ FBA311WT
Description
- Type I barricade measures 45 x 25 in.
- Reflective sheeting: 12 x 24 in. (on upper panel only)
- Engineer-Grade orange/white reflective stripe sheeting on two sides of white plastic barricades
- Molded-in handle with clock/lock hinges
- Folds flat for storage and transport
- Hollow legs can be filled with gravel or sand for added stability
Specifications
45 x 25 in. | |
Warning Sign | |
White Striped |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction