Learn More
Saint Gobain Performance Plastics Tube Tygon .050 ID 500FT-150M/PK
Supplier: Saint Gobain Performance Plastics AAD04133
Saint-Gobain Tygon ND 100-80 Tubing Diameter Inner 0.05 in. Diameter Outer 0.09 in. Thickness 0.02 in. Clear For Use With Medical grade