Learn More
DuPont™ Tychem™ 9000 Bib Overall and Jacket Combo
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Description
DuPont™ Tychem™ 9000 is strong and durable for consistent and lasting protection.
- NFPA 1994 Class 2 and NFPA 1992 certified
- Provides at least 30 minutes of protection against over 280 chemical challenges
- Used by many in the chemical handling and petrochemical manufacturing
Specifications
Color | Yellow |
Material | Santoprene |
Size | X-Large |
Ankle Style | Open ankles |
Seam Type | Taped |
Style | Double storm flap jacket |
Suspender | Adjustable webbing straps with buckle closure |