Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ Tychem™ 2000 SFR Apron
missing translation for 'orderingAttributeHoverText'
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
- Tychem™ 2000 SFR represents a new generation of secondary flame-resistant chemical garment technology
- Unlike traditional secondary flame resistant chemical garments that have been available for years, new Tychem™ 2000 SFR garments were specially designed to meet dual hazard needs of a protective chemical suit with secondary flame resistance
- This unique combination of performance permits Tychem™ 2000 SFR garments to be worn over primary flame resistant (FR) garments like Nomex™ when chemical splash and flash fire hazards exist
- The fabric used in Tychem™ 2000 SFR garments utilize a unique technology so that it does not char like traditional secondary flame resistant technologies - it is designed to shrink away from flame without burning
- Tychem™ 2000 SFR garments also provide an effective barrier against a range of inorganic acids, bases, industrial cleaning chemicals, and particles
- They're an ideal option for keeping workers safe from chemicals, while not compromising the flame protection of fire-resistance clothing worn beneath them
- Applications include: refineries, petrochemical plants, laboratories, hazardous maintenance operations
Specifications
Size | 3X-Large |
Color | Green |
Disposable | Single-use |
Closure Type | Hook and Loop, Waist Ties |
Sterility | Non-sterile |
Cuff Style | Elastic |
Latex Free | Yes |
Thickness | 7.5 mil |
Length (English) | 46.5 in. |
Length (English) Cuff | 33.75 in. |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction