Learn More
Supertek™ Two Potato Clock
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
This Two Potato Clock is designed to demonstrate the principle of an electrochemical call.
Supplier: Supertek™ CH12973S3
Description
- Watch the excitement!
- LCD clock
- Will it run on 2 potatoes - oranges - grapefruits or cans of soda pop?
- Ages 7 plus
Specifications
Potato Clock | |
LCD |