Learn More
Diversified Spaces™ Two-Book Compartment Oak Table with ChemGuard Top: Leg Brace, Stretcher and Caster Option
SCI_ED
Ideal for middle school or for use with milder chemicals. Diversified Spaces™ Oak Tables with ChemGuard Tops incorporates the decorative features of high pressure laminate into a durable, chemical resistant surfacing material.
$851.00 - $1315.00
Specifications
Certifications SEFA compliant and MAS Certified Green™
Finish UV
Frame Material Red Oak
Height (English) 33 in.
Height (English) Storage Area 3.875 in.
View More Specs
Products 16
Catalog Number Mfr. No. Depth (English) Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Depth (English) Width (English) Price Quantity & Availability  
S38199
SCI_ED
View Documents
Diversified Spaces™
C7102K30SC
24 in. 48 in.
Each for $851.00
Add to Cart
S38252
SCI_ED
View Documents
Diversified Spaces™
C7122K30SC
30 in. 48 in.
Each for $984.00
Add to Cart
S38281
SCI_ED
View Documents
Diversified Spaces™
C7132K30SC
30 in. 54 in.
Each for $1,053.00
Add to Cart
S38311
SCI_ED
View Documents
Diversified Spaces™
C7142K30SC
30 in. 60 in.
Each for $1,082.00
Add to Cart
S38342
SCI_ED
View Documents
Diversified Spaces™
C7152K30SC
30 in. 72 in.
Each for $1,213.00
Add to Cart
S38367
SCI_ED
View Documents
Diversified Spaces™
C7162K30SC
21 in. 48 in.
Each for $944.00
Add to Cart
S38396
SCI_ED
View Documents
Diversified Spaces™
C7172K30SC
36 in. 48 in.
Each for $1,119.00
Add to Cart
S38419
SCI_ED
View Documents
Diversified Spaces™
C7182K30SC
21 in. 54 in.
Each for $1,011.00
Add to Cart
S47047
SCI_ED
View Documents
Diversified Spaces™
C7192K30SC
36 in. 54 in.
Each for $1,210.00
Add to Cart
S47070
SCI_ED
View Documents
Diversified Spaces™
C7202K30SC
24 in. 54 in.
Each for $962.00
Add to Cart
S47103
SCI_ED
View Documents
Diversified Spaces™
C7212K30SC
21 in. 60 in.
Each for $1,026.00
Add to Cart
S47132
SCI_ED
View Documents
Diversified Spaces™
C7222K30SC
36 in. 60 in.
Each for $1,216.00
Add to Cart
S47152
SCI_ED
View Documents
Diversified Spaces™
C7232K30SC
21 in. 72 in.
Each for $1,107.00
Add to Cart
S47173
SCI_ED
View Documents
Diversified Spaces™
C7242K30SC
36 in. 72 in.
Each for $1,315.00
Add to Cart
S47190
SCI_ED
View Documents
Diversified Spaces™
C7302K30SC
24 in. 72 in.
Each for $1,092.00
Add to Cart
S47219
SCI_ED
View Documents
Diversified Spaces™
C7602K30SC
24 in. 60 in.
Each for $946.00
Add to Cart
Description
Description
  • 1.25 in. ChemGuard Table Surface
  • 2 book compartments measuring 14W x 13.625D x 3.875 in. H
  • Equipped with leg braces, stretcher and 3 in. locking swivel casters
  • 2.25 in. solid square legs, with 5/16 in. hanger bolts installed, supplied nuts and washers
  • Aprons are 13/16D x 4.5 in. H with heavy-duty 13 gauge steel corner braces
  • Equipped with non-skid adjustable glides for precise leveling and rubber leg boots
  • Finished natural with a chemical resistant UV finish
  • Solid Oak Construction
  • Minimal assembly required; legs need to be attached
  • Weight capacity is 500lbs evenly distributed
  • SEFA compliant and MAS Certified Green
  • Limited Lifetime Warranty
  • Made in the USA
Order Info
Leg braces and stretchers are recommended for use with tables 72 in. wide or more. Leg braces and stretchers sold separately.
Specifications
Specifications
SEFA compliant and MAS Certified Green™
Red Oak
3.875 in.
Red Oak
1 Ea.
3 in. locking swivel casters
Yes
500 lb.
226.8 kg
UV
33 in.
2-book compartments, leg braces, stretchers and casters
Leg brace, stretcher and casters
Lifetime
14 in.
1.25 in. ChemGuard
Book Compartment Table