Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eppendorf™ twin.tec™ real-time qPCR Plates, 96-well - qPCR Plates
Eppendorf twin.tec™ real-time PCR plates, designed for low-volume qPCR, utilize white wells to enhance fluorescent light reflection, improving sensitivity and reproducibility in assays for reliable results.
Supplier: Eppendorf™ 0030132718
Description
- White wells for better reflection.
- High mechanical stability.
- Effective sealing by raised well rims.
- Skirted and semi-skirted frames.
- Optimal heat transfer due to reduced wall thickness.
- Autoclavable (121°C, 20 min.).
- Available in lot-certified PCR clean and Eppendorf Forensic DNA Grade for tested purity.
Specifications
Includes Barcode | |
8 x 12 | |
20 Plates (5 Bags x 4 Plates) | |
PCR Plate | |
Polypropylene, Polycarbonate | |
96 | |
RT-PCR applications | |
PCR Clean (Non-sterile) | |
Yes |
Standard | |
Autoclavable | |
Blue | |
Unskirted | |
150 μL | |
Non-sterile | |
6000 x g | |
20 Pk. | |
White |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction