Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaMotte™ Turbidity Standards
For use with LaMotte Portable Turbidity Meters
Supplier: LaMotte™ 1454
Description
Packaged in 60mL bottles
Specifications
Turbidity Standard | |
10 FNU Standard (ISO), 60 mL | |
2.02 oz. |
2020WI Portable Turbidity Meter | |
ISO | |
60 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction