Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Turbidity Standards
SCI_ED

For use with LaMotte Portable Turbidity Meters

Supplier:  LaMotte™ 1451

 View more versions of this product

Catalog No. S72351


Add to Cart

Description

Description

Packaged in 60mL bottles

Specifications

Specifications

Turbidity Standard
10 NTU
2.02 oz.
2020WE Portable Turbidity Meter
EPA
60 mL
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit