Learn More
NSI Environmental Solutions Turbidity QC Check Standards 25 NTU, 500mL
Supplier: NSI Environmental Solutions QCI154
A formazin based CRM available as a 500mL bottle. Stable and ready to use as received. Do not freeze.