Learn More
United Scientific Supplies Tuning Fork Set of 13
Tuning Fork Set of 13 includes a storage tray.
Supplier: United Scientific Supplies TFSET13
This item is not returnable.
View return policy
Description
- Convenient assortments of tuning forks. Made of aluminum alloy.
- The frequency and scale letter are stamped on each fork.
- Set of 13 contains frequencies of 100, 128, 256, 288, 320, 341.3, 384, 426.6, 480, 512, 1024, 2048, and 4096 Hz.
Specifications
Tuning Forks | |
Aluminum Alloy |