Learn More
United Scientific Supplies Tuning Fork Box of 8
Tuning Fork Set of 8 includes a handy storage box.
Supplier: United Scientific Supplies TFBOX8
This item is not returnable.
View return policy
Description
- Tuning Fork Boxed Set of 8 includes a wooden box with foam lining.
- Each aluminum alloy tuning fork has a specific place in the foam lining die-cut for easy storage.
- Also contains a mallet specially designed with one hard and one soft end for use with all frequencies of forks.
- Contains one each of tuning forks with frequencies of 256, 288, 320, 341.3, 384, 426.6, 480, and 512 Hz.
Specifications
Tuning Forks | |
Aluminum Alloy |