Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dynalon™ Tuff Vials
Ideal for leakproof shipping of reagents and buffer solutions, for packaging of serum, veterinary, pharmaceuticals, cosmeceutical and nutraceutical extracts. Dynalon™ Tuff Vials are used with most aromatic compounds and essential oils.
Supplier: Dynalon™ 6264340010
Description
- Vials are made of break resistant polycarbonate
- Includes polypropylene leakproof closures
- Reusable and autoclavable
- Clear glass helps viewing of valuable contents, liquid or solid
- 180mL can also be used as an outer protective container
Specifications
Clear | |
No | |
For leakproof shipping of reagents and buffer solutions; for packaging of serum, veterinary, cosmeceutical and nutraceutical extracts | |
Autoclavable | |
20-415 |
Polycarbonate | |
10 mL | |
Polypropylene | |
Screw Cap |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction